missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | EGFR |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL0822 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P00533, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?