missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eg5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NB500-181SS
This item is not returnable.
View return policy
Description
Eg5 Polyclonal specifically detects Eg5 in Human, Mouse, Rat, Porcine, Drosophila, Mammal, Plant samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting.
Specifications
| Eg5 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500, Immunoblotting reported in scientific literature (PMID 28392145) | |
| P52732 | |
| KIF11 | |
| A recombinant segment of the coiled-coil domain of human Eg5 [Uniprot: P52732] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, rat and porcine. Predicted to react with most mammalian species. Immunogen sequence has 98% identity to primate. Drosophila reactivity reported in scientific literature (PMID: 23888285). Silvetia compressa reactivity reported in scientific literature (PMID: 19527496). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| Eg5, HKSP, kinesin family member 11, kinesin-like 1, Kinesin-like protein 1, kinesin-like protein KIF11, Kinesin-like spindle protein HKSP, Kinesin-related motor protein Eg5, KNSL1TRIP-5, thyroid receptor interacting protein 5, Thyroid receptor-interacting protein 5, TR-interacting protein 5, TRIP5EG5 | |
| Rabbit | |
| 125 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Cytoskeleton Markers, Signal Transduction, Stem Cell Markers | |
| 3832 | |
| Human, Mouse, Rat, Pig, Drosophila, Mammalia, Plant | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction