missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2K Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00€ - 474.00€
Specifications
| Antigen | EEF2K |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated |
| Applications | Western Blot, Immunofluorescence, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635401
|
Novus Biologicals
NBP2-92950-0.02ml |
0.02 mL |
226.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646871
|
Novus Biologicals
NBP2-92950-0.1ml |
0.1 mL |
474.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EEF2K Polyclonal antibody specifically detects EEF2K in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Gene Knock-OutSpecifications
| EEF2K | |
| Western Blot, Immunofluorescence, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia Signaling | |
| PBS with 50% glycerol, pH7.3. | |
| 29904 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase, calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase, EC 2.7.11, EC 2.7.11.20, eEF-2 kinase, eEF-2Kcalmodulin-dependent protein kinase III, elongation factor-2 kinase, eukaroytic elongation factor 2 kinase, eukaryotic elongation factor 2 kinase, eukaryotic elongation factor-2 kinase, HSU93850, MGC45041 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 406-725 of human EEF2K (NP_037434.1). KLDHLHWPVFSDLDNMASRDHDHLDNHRESENSGDSGYPSEKRGELDDPEPREHGHSYSNRKYESDEDSLGSSGRVCVEKWNLLNSSRLHLPRASAVALEVQRLNALDLEKKIGKSILGKVHLAMVRYHEGGRFCEKGEEWDQESAVFHLEHAANLGELEAIVGLGLMYSQLPHHILADVSLKETEENKTKGFDYLLKAAEAGDRQSMILVARAFDSGQNLSPDRCQDWLEALHWYNTALEMTDCDEGGEYDGMQDEPRYMMLAREAEMLFTGGYGLEKDPQRSGDLYTQAAEAAMEAMKGRLANQYYQKAEEAWAQMEE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title