missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ECE1 (Human) Recombinant Protein (P01)
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Human ECE1 full-length ORF ( NP_001388.1, 1 a.a. - 770 a.a.) recombinant protein with GST-tag at N-terminal
- Theoretical MW: 113.6kDa
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Best use within three months from the date of receipt of this protein
Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array
Specifications
Specifications
| Accession Number | NP_001388.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gene ID (Entrez) | 1889 |
| Molecular Weight (g/mol) | 113.6 |
| Name | ECE1 (Human) Recombinant Protein (P01) |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction