Learn More
Ebastine, Mikromol™
Product Code. 17958464
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958464 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17958464 Supplier Mikromol Supplier No. MM0468.000250
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ebastine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ebastine
CAS 90729-43-4
Analyte or Component Names Ebastine
Shipping Condition Room Temperature
Molecular Formula C32 H39 N O2
Certifications/Compliance ISO 17034
Synonym Ebastine, 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-piperidinyl]-1-butanone, Bastel, Ebastel, Ebastin, LAS-W 090, RP 64305
InChI Formula InChI=1S/C32H39NO2/c1-32(2,3)28-18-16-25(17-19-28)30(34)15-10-22-33-23-20-29(21-24-33)35-31(26-11-6-4-7-12-26)27-13-8-5-9-14-27/h4-9,11-14,16-19,29,31H,10,15,20-24H2,1-3H3
SMILES CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4
IUPAC Name 4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
Molecular Weight (g/mol) 469.66
Quantity 250 mg
Formula Weight 469.2981 g/mol