missing translation for 'onlineSavingsMsg'
Learn More

Methenolone 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30463783
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30463783

Brand: LoGiCal LGCAMP1374.0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methenolone
CAS 153-00-4
Analyte or Component Names Methenolone
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H30 O2
Concentration 1 mg/ml
Certifications/Compliance ISO 17034
Synonym Methyldopa Methyl Ester, (5alpha,17beta)-17-Hydroxy-1-methyl-androst-1-en-3-one, 5alpha-Androst-1-en-3-one, 17beta-hydroxy-1-methyl- (6CI,7CI,8CI), (5alpha,17beta)-17-Hydroxy-1-methylandrost-1-en-3-one, 1-Methyl-5alpha-androst-1-en-17beta-ol-3-one, 1-Methyl-Delta1-androsten-17beta-ol-3-one, 17beta-Hydroxy-1-methyl-5alpha-androst-1-en-3-one, Metenolone, Methenolon, Methenolone
Solution Type Single Solution
SMILES CC1=CC(=O)C[C@@H]2CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@@H]3[C@@]12C
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 302.43
Quantity 1 mL
Formula Weight 302.428 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.