Learn More
Thermo Scientific™ EASY-Column™ Capillary HPLC Column Connector Kits
Product Code. 11302150 Shop All Thermo Scientific Products
Click to view available options
Type:
A/B mixing and venting for two-column setup
HPLC to column
Kit for 360μm OD fused silica
Two-column setup
Unit Size:
Each
Pack of 30
Product Code. 11302150
Brand: Thermo Scientific™ SC600
Only null left
Add to basket
Add to basket
Use these connectors, tees and sleeves with Thermo Scientific™ EASY-Column™ Capillary HPLC Columns.
Use these connector kits, sleeves, and tees with Thermo Scientific™ EASY-Column™ Capillary Columns. EASY-Columns optimize nanoscale LC and LC/MS with. Easy-to-use, zero dead volume connector kits are available for one- and two-column configurations and ensure simple, reproducible setup for on-line LC/MS. Each column delivers excellent chromatographic performance on any nanoscale LC system. At flow rates of nL/min versus mL/min, the EASY-Column offers higher sensitivity with greater signal-to-noise ratio compared to traditional electrospray.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.79 mm
For Use With LC/MS
Pore Size 120 Å
Product Line EASY-Column
Endcapped Yes
Type HPLC to column