missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam E3 Ligase Auto-Ubiquitylation Assay Kit, Enzyme Activity, 10 Test
Description
- Assay type: Enzyme activity
- Sample types: Purified protein
- Citation Count: 10
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Western Blot Kit |
| Format | Kit |
| No. of Reactions | 10 Tests |
| Target Molecule | E3 Ligase |
| Detection Method | Colorimetric |
| Sample Type | Purified protein |
| Quantity | 10 Tests |
| For Use With (Application) | E3 Ligase Auto-Ubiquitylation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?