Learn More
E2F7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92216-0.1ml
This item is not returnable.
View return policy
Description
E2F7 Polyclonal antibody specifically detects E2F7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| E2F7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E2F transcription factor 7, E2F-7, FLJ12981, transcription factor E2F7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 540-740 of human E2F7 (NP_976328.2). LLAGQPLVYVPSASLFMLYGSLQEGPASGSGSERDDRSSEAPATVELSSAPSAQKRLCEERKPQEEDEPATKRQSREYEDGPLSLVMPKKPSDSTDLASPKTMGNRASIPLKDIHVNGQLPAAEEISGKATANSLVSSEWGNPSRNTDVEKPSKENESTKEPSLLQYLCVQSPAGLNGFNVLLSGSQTPPTVGPSSGQLPS | |
| 0.1 mL | |
| Signal Transduction | |
| 144455 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |