Learn More
E2F-1 Antibody (1D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26609-50ul
 View more versions of this product
Product Code. 30043235
  • 343.00€ / 50µL
Description
E2F-1 Monoclonal antibody specifically detects E2F-1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E2F-1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
1D12
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1
A synthesized peptide derived from Human E2F-1 [UniProt Q01094]
50 μL
Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators
1869
Store at -20 to -70C. Avoid freeze-thaw cycles.