Learn More
(E)-Mevinphos (cis-butenoic acid) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315067
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315067 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315067 Supplier Dr. Ehrenstorfer Supplier No. DREA15221000AL100
 Restricted Item
Estimated Shipment: 09-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material E-Mevinphos (cis-butenoic acid)
CAS 298-01-1
Analyte or Component Names E-Mevinphos (cis-butenoic acid)
Shipping Condition Room Temperature
Molecular Formula C7 H13 O6 P
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2-Butenoic acid, 3-[(dimethoxyphosphinyl)oxy]-, methyl ester, (E)-, Crotonic acid, 3-hydroxy-, methyl ester, dimethyl phosphate, (E)- (8CI), (E)-Mevinphos, OS 2046, cis-Mevinphos, cis-Phosdrin, α-Mevinphos
InChI Formula InChI=1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3/b6-5+
SMILES COC(=O)\C=C(/C)\OP(=O)(OC)OC
IUPAC Name methyl (E)-3-dimethoxyphosphoryloxybut-2-enoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 224.15
Quantity 1 mL
Formula Weight 224.045 g/mol