Learn More
E. Coli 015 Antibody (15/443), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64541AF350
486.20 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
E. Coli 015 Monoclonal antibody specifically detects E. Coli 015 in Bacteria samples. It is validated for ELISA, Immunofluorescence
Specifications
| E. Coli 015 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA, Immunofluorescence | |
| 15/443 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Escherichia coli | |
| Native, NCTC 9015 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 |