missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ dystrophia myotonica containing WD repeat motif Antibody [DyLight 488], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30748G
This item is not returnable.
View return policy
Description
dystrophia myotonica containing WD repeat motif Polyclonal antibody specifically detects dystrophia myotonica containing WD repeat motif in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| dystrophia myotonica containing WD repeat motif | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| D19S593E, DM9, DMRN9, DMR-N9, dystrophia myotonica WD repeat-containing protein, dystrophia myotonica, WD repeat containing, dystrophia myotonica-containing WD repeat motif, Dystrophia myotonica-containing WD repeat motif protein, gene59, Protein 59, Protein DMR-N9 | |
| Produced in rabbits immunized with E. coli-derived Human dystrophia myotonica containing WD repeat motif fragment | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1762 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction