Description
Thermo Scientific™ DyNAzyme™ EXT DNA Polymerase is a versatile and easy-to-use enzyme especially suited for handling demanding reaction conditions without time-consuming optimization
It is an optimized mixture of DyNAzyme II DNA Polymerase and a proofreading PCR enzyme. DyNAzyme EXT DNA Polymerase offers exceptional performance that Taq-based mixtures cannot match.
- Ideal for difficult templates, such as GC rich and looped sequences
- Improved fidelity compared to standard PCR enzymes
- Generates fragments up to 40 kb in long PCR reactions
- High yields, robust reactions with standard templates
- More thermostable than Taq DNA polymerase
Recommended for:
Difficult templates, long-range PCR, DHPLC, microarray, cloning
Specifikationer
Specifikationer
| Content And Storage | 10x Optimized DyNAzyme EXT Buffer, 10x Mg2+-free DyNAzyme EXT Buffer, 50 mM MgCl2 -solution, DMSO. Optimized DyNAzyme EXT Buffer provides 1.5 mM MgCl2 in the final 1x concentration. Recommended storage temperature -20°C. Stable for one year from the assay date. |
| Description | DyNAzyme EXT DNA Polymerase |
| Quantity | 200U (1U/µL) |
| For Use With (Application) | Microarray Analysis |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?