Learn More
DYKDDDDK Mouse anti-Recombinant Protein, CoraLite 594, Clone: 1E7B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66008-100UL
Description
The DYKDDDDK antibody from Proteintech is a mouse monoclonal antibody generated with peptide of DYKDDDDK. This antibody recognizes recombinant protein antigen. The DYKDDDDK antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| DYKDDDDK | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| IgG2b |
| Immunofluorescence | |
| 1E7B4 | |
| Mouse | |
| Primary |