Learn More
Dutch Seven PCB Mixture 317 1 μg/mL in Isooctane, Dr. Ehrenstorfer
Product Code. 30327144
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327144 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327144 Supplier Dr. Ehrenstorfer Supplier No. DREA50000317IO
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
CAS 37680-73-2, 31508-00-6, 35065-28-2, 35065-27-1, 35065-29-3, 7012-37-5
Quantity 1 mL
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Isooctane
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.