missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUOX2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-61576AF647
This item is not returnable.
View return policy
Description
DUOX2 Polyclonal specifically detects DUOX2 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| DUOX2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
| Q9NRD8 | |
| DUOX2 | |
| Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-Prot# Q9NRD8] | |
| 0.1 mL | |
| Cell Biology, Lipid and Metabolism, Neuroscience | |
| 50506 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| dual oxidase 2, dual oxidase-like domains 2, EC 1.11.1.-, EC 1.6.3.1, flavoprotein NADPH oxidase, Large NOX 2, LNOX2P138(TOX), Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF2, p138 thyroid oxidase, P138-TOX, THOX2TDH6, Thyroid oxidase 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction