missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dulbecco's Phosphate Buffered Saline with 2% Fetal Bovine Serum

Product Code. 17191036
Encompass_Preferred
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17191036

Brand: STEMCELL Technologies 07905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell culture buffer

  • Dulbecco's Phosphate Buffered Saline with 2% Fetal Bovine Serum is recommended for use as a buffer to dilute or wash nucleated cells

Specifications

Quantity 500 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.