missing translation for 'onlineSavingsMsg'
Learn More

DTX4 Antibody, HRP, Novus Biologicals™

Product Code. 18608611 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18608611 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18608611 Supplier Novus Biologicals Supplier No. NBP281924H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DTX4 Polyclonal antibody specifically detects DTX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Deltex 4 Homolog, Deltex 4 Homolog (Drosophila), Deltex Homolog 4 (Drosophila), deltex4, E3 Ubiquitin-Protein Ligase DTX4, EC 6.3.2., KIAA0937, Protein Deltex-4, RING Finger Protein 155, RNF155
Host Species Rabbit
Immunogen DTX4 antibody was raised against a 19 amino acid peptide near the center of human DTX4. The immunogen is located within amino acids 230 - 280 of DTX4. Amino Acid Squence: AKPLDSTGTIRGPLKTAPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23220
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.