missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81924
This item is not returnable.
View return policy
Description
DTX4 Polyclonal antibody specifically detects DTX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| DTX4 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| DTX4 | |
| DTX4 antibody was raised against a 19 amino acid peptide near the center of human DTX4. The immunogen is located within amino acids 230 - 280 of DTX4. Amino Acid Squence: AKPLDSTGTIRGPLKTAPS | |
| Affinity Purified | |
| RUO | |
| 23220 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| Deltex 4 Homolog, Deltex 4 Homolog (Drosophila), Deltex Homolog 4 (Drosophila), deltex4, E3 Ubiquitin-Protein Ligase DTX4, EC 6.3.2., KIAA0937, Protein Deltex-4, RING Finger Protein 155, RNF155 | |
| Rabbit | |
| 68 kDa | |
| 0.1 mg | |
| Primary | |
| DTX4 antibody is human and mouse reactive. At least two isoforms of DTX4 are known to exist; this antibody will detect both isoforms. DTX4 antibody is predicted to not cross-react with other DTX protein family members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction