missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Antibody (J2) [Janelia Fluor™ 585], Novus Biologicals™

Product Code. 30163288
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30163288 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30163288 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP311395JF585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Dot Blot,Affinity Purification,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen dsRNA
Applications Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Dot Blot, Affinity Purification, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone J2
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen Female DBA/2 mice were injected intraperitonially with a mixture of 50 μg L-dsRNA and 75 μg methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.