missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsDNA Rabbit anti-Human, Clone: DSD/4054R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07302-100ug
This item is not returnable.
View return policy
Description
dsDNA Monoclonal specifically detects dsDNA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| dsDNA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Rabbit | |
| Bioreactor concentrate | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DSD/4054R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Nuclei of Burkitt's cells | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction