missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DSCC1 Monoclonal specifically detects DSCC1 in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
Specifications
| Antigen | DSCC1 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | V76P1B2*B12 |
| Conjugate | FITC |
| Dilution | ELISA, Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1 |
| Gene Symbols | DSCC1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?