Learn More
Epredia™ Dry Powder Stains
Product Code. 10434720 Shop All Epredia Products
Click to view available options
Type:
Eosin
Unit Size:
100g
Product Code. 10434720
Brand: Epredia™ 90017
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Stain specimens with Eosin-Y or Wright compounds with Epredia™ Dry Powder Stains, certified by the Biological Stain Commission.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Type Eosin
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acid red 87