missing translation for 'onlineSavingsMsg'
Learn More

DRAP1 Antibody (PCRP-DRAP1-1A12), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18650250 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18650250 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18650250 Supplier Novus Biologicals Supplier No. NBP324152AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DRAP1 Monoclonal antibody specifically detects DRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAP1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-DRAP1-1A12
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias dr1-associated corepressor, Dr1-associated protein 1, DR1-associated protein 1 (negative cofactor 2 alpha), NC2-alphaNegative co-factor 2-alpha, negative cofactor 2 alpha
Host Species Mouse
Immunogen Recombinant human DRAP1 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10589
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.