missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DPPIV/CD26 Antibody (begelomab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | DPPIV/CD26 |
|---|---|
| Clone | begelomab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044396
|
Novus Biologicals™
NBP3-28493-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047634
|
Novus Biologicals™
NBP3-28493-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047181
|
Novus Biologicals™
NBP3-28493-100ug |
100 μg |
337.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| DPPIV/CD26 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| DPP4/ CD26 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| begelomab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P27487 | |
| 1803 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title