missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP2 Monoclonal Antibody (398024)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561305
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
Specifications
| DPP2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UHL4 | |
| DPP7 | |
| Mouse myeloma cell line NS0-derived recombinant human DPPII/QPP/DPP7, Gly22-Leu492. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 398024 | |
| Unconjugated | |
| DPP7 | |
| carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase | |
| Rat | |
| Protein A/G | |
| RUO | |
| 29952 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction