missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DPEP3 Antibody (tamrintamab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28580-50ug
This item is not returnable.
View return policy
Description
DPEP3 Monoclonal antibody specifically detects DPEP3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DPEP3 | |
| Monoclonal | |
| Unconjugated | |
| Q9H4B8 | |
| Human | |
| Protein A purified | |
| RUO | |
| 64180 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| tamrintamab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Dipeptidase 3, EC 3.4.13.19, MBD3, Membrane-Bound Dipeptidase 3, Metallopeptidase (Family M19) | |
| DPEP3 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction