missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPEP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92470-0.02ml
This item is not returnable.
View return policy
Description
DPEP1 Polyclonal antibody specifically detects DPEP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DPEP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dehydropeptidase-I, dipeptidase 1, dipeptidase 1 (renal), EC 3.4.13.19, hRDP, MDPMBD1, RDPMicrosomal dipeptidase, Renal dipeptidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-385 of human DPEP1 (NP_001121613.1). DFFRDEAERIMRDSPVIDGHNDLPWQLLDMFNNRLQDERANLTTLAGTHTNIPKLRAGFVGGQFWSVYTPCDTQNKDAVRRTLEQMDVVHRMCRMYPETFLYVTSSAGIRQAFREGKVASLIGVEGGHSIDSSLGVLRALYQLGMRYLTLTHSCNTPWADNWLVDTGDSEPQSQGLSPFGQRVVKELNRLGVLIDLAHVSVATMKATLQLSRAPVIFSHSSAYSVCASRRNVPDDVLRLVKQTDSLVMVNFYNNYISCTNKANLSQVADHLDHIKEVAGARAVGFGGDFDGVPRVPEGLEDVSKYPDLIAELLRRNWTEAEVKGALADNLLRVFEAVEQASNLTQAPEEEPIPLDQLGGSCRTHYGYSS | |
| 0.02 mL | |
| Cancer, Cell Biology | |
| 1800 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction