missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPCR1 Monoclonal Antibody (986124)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561300
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The DPCR1 gene, known as diffuse panbronchiolitis critical region 1, encodes a protein involved in immune responses and epithelial cell function, particularly in the respiratory system. Although the specific functions of DPCR1 are not extensively characterized, the gene is located within a region linked to diffuse panbronchiolitis, a chronic inflammatory lung disease. DPCR1 is thought to contribute to immune regulation, potentially influencing innate immune responses and epithelial cell interactions critical for maintaining respiratory health. Its expression in airway epithelial cells suggests a role in modulating mucus production, barrier integrity, and protection against pathogens. Aberrant expression of DPCR1 may impact respiratory function, contributing to conditions such as asthma or chronic obstructive pulmonary disease (COPD) by affecting immune responses and inflammation. Understanding DPCR1's functions and regulatory mechanisms in the respiratory system is essential for exploring its potential as a therapeutic target for respiratory diseases characterized by chronic inflammation and immune dysregulation.
Specifications
| DPCR1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q3MIW9 | |
| DPCR1 | |
| Human DPCR-1 synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 986124 | |
| Unconjugated | |
| DPCR1 | |
| bCX105N19.6; C6orf37; diffuse panbronchiolitis critical region 1; diffuse panbronchiolitis critical region protein 1; DPCR1; MGC126710; MGC126712; Mucin-like protein 3; MUCL3; PBLT | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 135656 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction