missing translation for 'onlineSavingsMsg'
Learn More

Abcam Doxycycline Hyclate, Matrix Metalloprotease Inhibitor, Solid, 25 g

Product Code. 32120098
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120098 Supplier Abcam Supplier No. ab14109125g
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic tetracycline and matrix metalloprotease inhibitor that inhibits tumour growth, suppresses cell proliferation, and induces apoptosis.

  • Chemical Type: Synthetic tetracycline
  • Biological Activity: Matrix metalloprotease (MMP) inhibitor; inhibits tumour growth, suppresses cell proliferation, and induces apoptosis in vitro and in vivo
  • Purity: >97%
  • Molecular Weight: 1,025.9 Da
  • Storage Temperature: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Matrix Metalloproteinase
Formulation 100% Doxycycline hyclate
Quantity 25 g
For Use With (Application) Matrix metalloprotease inhibition, tumour growth inhibition, cell proliferation suppression, apoptosis induction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.