Learn More
Doxazosin Mesilate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Doxazosin mesilate |
| CAS | 77883-43-3 |
| Analyte or Component Names | Doxazosin mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H25 N5 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxazosin Mesilate, Methanone, [4-(4-amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl](2,3-dihydro-1,4-benzodioxin-2-yl)-, methanesulfonate (1:1), Piperazine, 1-(4-amino-6,7-dimethoxy-2-quinazolinyl)-4-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]-, monomethanesulfonate (9CI), 1,4-Benzodioxin, piperazine deriv., Alfadil, Cardenalin, Cardran, Cardular, Cardura, Cardura (pharmaceutical), Carduran, Diblocin, Doxazosin mesylate, Doxazosin monomethanesulfonate, Normothen, Supressin, UK 33274-27, doxazolazine mesylate |
| InChI Formula | InChI=1S/C23H25N5O5.CH4O3S/c1-30-18-11-14-15(12-19(18)31-2)25-23(26-21(14)24)28-9-7-27(8-10-28)22(29)20-13-32-16-5-3-4-6-17(16)33-20;1-5(2,3)4/h3-6,11-12,20H,7-10,13H2,1-2H3,(H2,24,25,26);1H3,(H,2,3,4) |
| Solution Type | Neat |
| SMILES | COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4COc5ccccc5O4.CS(=O)(=O)O |
| Show More |