missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOT1L Monoclonal Antibody (OTI8A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802739S
This item is not returnable.
View return policy
Description
DOT1L Monoclonal antibody specifically detects DOT1L in Human samples. It is validated for Western BlotSpecifications
| DOT1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TEK3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8A11 | |
| Unconjugated | |
| Dot1l | |
| A630076O07; AW907654; DKFZp586P1823; DOT1; DOT1 like histone H3K79 methyltransferase; DOT1 like histone lysine methyltransferase; DOT1L; DOT1-like histone H3 methyltransferase; DOT1-like histone H3K79 methyltransferase; DOT1-like protein; DOT1-like, histone H3 methyltransferase; DOT1-like, histone H3 methyltransferase (S. cerevisiae); H3-K79-HMTase; histone H3 methyltransferase DOT1; histone H3-K79 methyltransferase; histone methyltransferase DOT1L; histone-lysine N-methyltransferase, H3 lysine-79 specific; KIAA1814; KMT4; lysine N-methyltransferase 4; mDot1 | |
| Human recombinant protein fragment corresponding to amino acids 1008-1424 of human DOT1L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction