missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOT1L Monoclonal Antibody (OTI8A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802739
This item is not returnable.
View return policy
Description
DOT1L encodes histone methyltransferase, and methylates ′Lys-79′ of histone H3. Nucleosomes are preferred as substrate compared to free histones, binds to DNA.Specifications
| DOT1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TEK3 | |
| DOT1L | |
| Human recombit protein fragment corresponding to amino acids 1008-1424 of human DOT1L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8A11 | |
| Unconjugated | |
| DOT1L | |
| DOT1, KMT4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction