missing translation for 'onlineSavingsMsg'
Learn More

DORFIN Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. p-200062178 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18607061 0.02 mL 0.02mL
18621361 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18607061 Supplier Novus Biologicals Supplier No. NBP2920510.02ml

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DORFIN Polyclonal antibody specifically detects DORFIN in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DORFIN
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp566B1346, dorfin, Double ring-finger protein, E3 ubiquitin-protein ligase RNF19A, EC 6.3.2, EC 6.3.2.-, p38, protein p38 interacting with transcription factor Sp1, ring finger protein 19, RING finger protein 19 isoform, ring finger protein 19Ap38 protein, ring-IBR-ring domain containing protein Dorfin, RNF19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 639-838 of human RNF19A (NP_056250.3). GSATRSHAGGSSSGLPEGKSSATKWSKEATAGKKSKSGKLRKKGNMKINETREDMDAQLLEQQSTNSSEFEAPSLSDSMPSVADSHSSHFSEFSCSDLESMKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTEI
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25897
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.