missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopa Decarboxylase/DDC Antibody (CL2962) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46642AF594
This item is not returnable.
View return policy
Description
Dopa Decarboxylase/DDC Monoclonal antibody specifically detects Dopa Decarboxylase/DDC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Dopa Decarboxylase/DDC | |
| Monoclonal | |
| Alexa Fluor 594 | |
| AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28 | |
| This antibody was developed using a recombinant protein derived from P20711, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 1644 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2962 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction