missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey F(ab) anti-Goat IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NB120-6667
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:200000, Immunohistochemistry 1:500 - 1:2500 | |
| Donkey F(ab) anti-Goat IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with goat whole molecule in donkey. | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| No reaction was observed against anti-Papain or anti-Donkey IgG F(c). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Goat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction