missing translation for 'onlineSavingsMsg'
Learn More

Donkey F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™

Product Code. 18244352 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18244352 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18244352 Supplier Novus Biologicals Supplier No. NB7596
  • ONLINE EXCLUSIVE PRICE
    591.00€
    558.60€ / 0.5mg
    Save 32.40€
    5% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Dot Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate PE
Dilution Western Blot 1:100-1:500, Flow Cytometry 1:100 - 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunohistochemistry-Paraffin 1:200-1:500, Dot Blot
Host Species Donkey
Immunogen Donkey F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in donkey.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Donkey IgG F(c), Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat or Sheep Serum Proteins.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.