missing translation for 'onlineSavingsMsg'
Learn More

Donkey F(ab')2 anti-Human IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™

Product Code. 18736333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18736333 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18736333 Supplier Novus Biologicals Supplier No. NB1207005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate PE
Dilution Flow Cytometry 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:250
Host Species Donkey
Immunogen Human IgG whole molecule
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Donkey IgG F(c), Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat or Sheep Serum Proteins.
Reconstitution Reconstitute to desired concentration.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.