missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey Anti-Sheep IgG NorthernLights™ NL557-conjugated Antibody, R&D Systems™
Donkey Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems NL010
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgG | |
| Polyclonal | |
| NorthernLights 557 (NL557) | |
| Immunoglobulin G, ImmunoglobulinG | |
| Donkey | |
| 0.5 mL | |
| Secondary | |
| Sheep | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| 1mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| IGHG1 | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction