missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody, DyLight 633 (Pre-adsorbed), Novus Biologicals™

Product Code. 18268497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18268497 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18268497 Supplier Novus Biologicals Supplier No. NBP175679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Flow Cytometry, Immunomicroscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 633
Dilution Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Host Species Donkey
Immunogen Purified Sheep IgG (H&L)
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on sheep IgG and light chains on all sheep immunoglobulins. This antibody has been pre-adsorbed against human, mouse, and rabbit IgG.
Reconstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Sheep
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.