missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Sheep IgG (H+L) Secondary Antibody, DyLight 488 (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-75446
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Sheep samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunomicroscopy
Specifications
| IgG (H+L) | |
| Polyclonal | |
| DyLight 488 | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free | |
| This Donkey anti-Sheep IgG (H+L) Secondary Antibody [DyLight 488] (Pre-adsorbed) was developed against purified sheep IgG (H&L). | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. | |
| IgG |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000 | |
| Donkey | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction