missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, DyLight 650, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-72698C
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rat IgG whole molecule | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| DyLight 650 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction