missing translation for 'onlineSavingsMsg'
Learn More

Donkey Anti-Rat IgG H&L (Alexa Fluor™ 750) preadsorbed, Abcam

Product Code. 32116333
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116333 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116333 Supplier Abcam Supplier No. ab175750500ug
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody

Suitable for WB. Preadsorbed to minimise non-specific binding and high background staining. Cited in 2 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ig gamma-1 chain C region (IGHG1)
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Ig gamma-1 chain C region
Gene Accession No. P20759
Host Species Donkey
Immunogen Ig gamma-1 chain C region (IGHG1)
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype Not Available
Shipping Condition Blue Ice
Target Gene Ig gamma-1 chain C region
Target Molecule Ig gamma-1 chain C region (IGHG1)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.