missing translation for 'onlineSavingsMsg'
Learn More

Donkey Anti-Rabbit IgG NorthernLights™ NL557-conjugated Antibody, R&D Systems™

Product Code. 18184461
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18184461 0.5 mL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18184461 Supplier R&D Systems Supplier No. NL004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody has been used in 16 publications

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 1mg/mL
Conjugate NorthernLights 557 (NL557)
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Rabbit
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.