missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody [PE], Novus Biologicals™
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Host Species | Donkey |
| Immunogen | This Donkey anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rabbit IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?