missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-75286PECY7
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunodiffusion, Immunoassay
Specifications
| IgG (H+L) | |
| Polyclonal | |
| PBS | |
| This Donkey anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rabbit IgG (H&L). | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| PE-Cyanine7 | |
| Donkey | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction