Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-72580
263.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:500 - 1:2500, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:200 - 1:1000 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry | |
| Alkaline Phosphatase (AP) | |
| 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Rabbit IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Rabbit | |
| IgG |