missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Concentration | 4.5 mg/mL |
| Conjugate | Unconjugated |
| Host Species | Donkey |
| Immunogen | Purified Mouse IgG (H&L) |
| Purification Method | Affinity purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?