missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Mouse IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™

Product Code. 18513422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18513422 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18513422 Supplier Novus Biologicals Supplier No. NBP175610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 350
Dilution Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Host Species Donkey
Immunogen This Donkey anti-Mouse IgG (H+L) Secondary Antibody [DyLight 350] was developed against purified mouse IgG (H&L).
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Reconstitution Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.